Natriummethanolaat for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Sodium methylate, Sodium methanolate

M-Clarity™ quality level = MQ200

8.06538.0005 8.06538.0250 8.06538.0500
8.06538.0005EA 31.8 EUR
8.06538.0005 8.06538.0250 8.06538.0500
Natriummethanolaat for synthesis, Sigma-Aldrich®
Natriummethanolaat
Formule: CH₃NaO
Smeltpunt: 127 °C
Dichtheid: 1,3 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012179
CAS nummer: 124-41-4
EINECS: 204-699-5
UN: 1431
ADR: 4.2 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 127 °C (decomposition)
Molar Mass 54.02 g/mol
Bulk Density 500 - 600 kg/m3
Boiling Point 300 °C
Vapor Pressure (20 °C)
Density 1.3 g/cm3 (20 °C)
Ignition Point 240 °C
Synonyms Sodium methoxide, Sodium methanolate
Hill Formula CH3NaO
Chemical Formula CH3ONa

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR