1,3-Propanediol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.07481.0250 8.07481.1000
8.07481.0250EA 82.7 EUR
8.07481.0250 8.07481.1000
1,3-Propanediol for synthesis, Sigma-Aldrich®
1,3-Propanediol
Formule: HO(CH₂)₃OH
Kookpunt: 214 °C (1013 hPa)
Smeltpunt: –32 °C
Dichtheid: 1,0597 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002949
CAS nummer: 504-63-2
EINECS: 207-997-3
Merck Index: 12,09845

Order Now

Specification Test Results

Solubility 100 g/l (20 °C)
Melting Point -32 °C
Molar Mass 76.09 g/mol
Boiling Point 213 - 215 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Refractive Index 1.4396 (20 °C, 589 nm)
Explosion Limit 2.5 %(V)
Density 1.05 g/cm3 (20 °C)
pH 4.5 - 7.0 (100 g/l, H2O, 20 °C)
Ignition Point 405 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR