1H-Pyrazole for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.07572.0025
8.07572.0025EA 78.5 EUR
8.07572.0025
1H-Pyrazole for synthesis, Sigma-Aldrich®
1H-Pyrazole
Formule: C₃H₄N₂
Kookpunt: 186 °C (1013 hPa)
Smeltpunt: 66…68 °C
Dichtheid: 1,34 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005234
CAS nummer: 288-13-1
EINECS: 206-017-1
Merck Index: 12,08144

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 66 - 68 °C
Molar Mass 68.07 g/mol
Boiling Point 186 - 188 °C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR