Sarcosine for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.07666.0250 8.07666.9010
8.07666.0250EA 104 EUR
8.07666.0250 8.07666.9010
Sarcosine for synthesis, Sigma-Aldrich®
Sarcosine
Formule: C₃H₇NO₂
Kookpunt: 185 °C (1013 hPa)
Smeltpunt: 63 °C
Dichtheid: 1,301 g/cm³ (20 °C)
MDL Nummer: MFCD00004279
CAS nummer: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519

Order Now

Specification Test Results

Solubility 1481 g/l (20°C)
Melting Point 208 °C
Molar Mass 600 kg/m3
Bulk Density 600 kg/m3
pH 6.1 (100 g/l, H2O, 20 °C)
Synonyms Methylaminoacetic acid, N-Methylglycine
Hill Formula C3H7NO2
Chemical Formula CH3NHCH2COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR