Trifluoroacetic anhydride ≥99% (by GC) for synthesis

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Trifluoracetic anhydride, TFAA

M-Clarity™ quality level = MQ200

8.08261.0501
8.08261.0501EA 441 EUR
8.08261.0501
Trifluoroacetic anhydride ≥99% (by GC) for synthesis
Trifluoroacetic anhydride
Formule: C₄F₆O₃
Kookpunt: 39,4 °C (1013 hPa)
Smeltpunt: –65 °C
Dichtheid: 1,487 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000416
CAS nummer: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8,I

Order Now

Specification Test Results

Assay (GC, area%) ≥99.0% (a/a)
Identity (IR) Passes Test
Density (d 20 °C/ 4 °C) 1.509 - 1.512


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR