4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221) ≥98% for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: [2.2.1]Cryptand, Kryptofix® 221, 4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane

M-Clarity™ quality level = MQ200

8.10646.0001 8.10646.0050 8.10646.8250
8.10646.0001EA 245 EUR
8.10646.0001 8.10646.0050 8.10646.8250
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221) ≥98% for synthesis, Sigma-Aldrich®
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221)
Formule: C₁₆H₃₂N₂O₅
MW: 332.44 g/mol
Dichtheid: 1.11 g/cm³ (20 °C)
MDL Nummer: MFCD00005108
CAS nummer: 31364-42-8
EINECS: 250-592-1

Order Now

Specification Test Results

Purity (GC) ≥ 98
Identity (IR-spectrum) passes test
Density (d 20 °C/ 4 °C) 1.107 - 1.114
Water (according to Karl Fischer) ≤ 0.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR