Antimony(V) fluoride zS, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


MQ200

8.12034.0100
8.12034.0100EA 850 EUR
8.12034.0100
Antimony(V) fluoride zS, Sigma-Aldrich®
Antimony(V) fluoride
Formule: F₅Sb
MW: 216.75 g/mol
Kookpunt: 149.4
Smeltpunt: 7
Dichtheid: 2.99 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011219
CAS nummer: 7783-70-2
EINECS: 232-021-8
UN: 1732
ADR: 8 (6.1) II
Merck Index: 13,00702

Order Now

Specification Test Results

Hill Formula F₅Sb
Chemical formula SbF₅
Molar Mass 216.74 g/mol
Boiling point 149 - 150 °C (1013 hPa)
Density 2.99 g/cm³ (20 °C)
Melting Point 7 °C
Solubility (20 °C) (rigorous decomposition)
Storage class 8B Non-combustible, corrosive hazardous materials
WGK WGK 2 water endangering
R Phrase R 14-20/22-34-51/53
S Phrase S 22-26-36/37/39-45-61

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR