Sodium bromate for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.14368.0250 8.14368.9025 8.14368.1000 8.14368.7500
8.14368.0250EA 47.3 EUR
8.14368.0250 8.14368.9025 8.14368.1000 8.14368.7500
Sodium bromate for synthesis, Sigma-Aldrich®
Sodium bromate
Formule: BrNaO₃
MW: 150.89 g/mol
Kookpunt: 1390
Smeltpunt: 381
Dichtheid: 3.254 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003476
CAS nummer: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1 II
Merck Index: 12,08736

Order Now

Specification Test Results

Solubility 364 g/l (20 °C)
Melting Point 381 °C (decomposition)
Molar Mass 150.89 g/mol
Density 3.3 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral,Aqueous solution
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR