1-Octadecanethiol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.14444.0050 8.14444.0250 8.14444.8000 8.14444.0010
8.14444.0050EA 33.7 EUR
8.14444.0050 8.14444.0250 8.14444.8000 8.14444.0010
1-Octadecanethiol for synthesis, Sigma-Aldrich®
1-Octadecanethiol
Formule: C₁₈H₃₈S
Kookpunt: 204 °C (1013 hPa)
Smeltpunt: 28…31 °C
Dichtheid: 0,8475 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004886
CAS nummer: 2885-00-9
EINECS: 220-744-1
UN: 3335
ADR: 9,

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 28 - 31 °C
Molar Mass 286.55 g/mol
Boiling Point 366 °C
Vapor Pressure  - 0.5 hPa (20 °C)
Flash Point 185 °C
Refractive Index 1.4645 (20 °C, 589 nm)
Density 0.847 g/cm3 (20 °C)
Ignition Point 200 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR