2-Chloroethanesulphonic acid sodium salt hydrate for synthesis

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium-2-chloroethanesulphonate hydrate

M-Clarity™ quality level = MQ200

8.14501.0050
8.14501.0050EA 196 EUR
8.14501.0050
2-Chloroethanesulphonic acid sodium salt hydrate for synthesis
2-Chloroethanesulphonic acid sodium salt hydrate
Formule: C₂H₄ClNaO₃S.H₂O
MDL Nummer: MFCD00007531
CAS nummer: 1089719-72-1
EINECS: 241-935-6

Order Now

Specification Test Results

Assay (acidimetric, calc. on anhydrous substance) ≥ 98.0 %
Water (K. F.) 9.0 - 11.0 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR