2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Diethylene glycol monoethyl ether acetate

M-Clarity™ quality level = MQ200

8.14635.1000 8.14635.9033
8.14635.1000EA 102 EUR
8.14635.1000 8.14635.9033
2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
2-(2-Ethoxyethoxy)ethyl acetate
Formule: C₈H₁₆O₄
MW: 176.21 g/mol
Kookpunt: 210
Smeltpunt: –25 °C
Dichtheid: 1.0114 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041928
CAS nummer: 112-15-2
EINECS: 203-940-1

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -25 °C
Molar Mass 176.21 g/mol
Boiling Point 217 - 218 °C
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 100 °C
Explosion Limit 1.5 %(V)
Density 1.01 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 310 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR