Glycolic acid ≥70%, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.14662.1000 8.14662.5000 8.14662.9026
8.14662.1000EA 142 EUR
8.14662.1000 8.14662.5000 8.14662.9026
Glycolic acid ≥70%, Sigma-Aldrich®
Glycolic acid
Formule: C₂H₄O₃
Dichtheid: 1,262 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004312
CAS nummer: 79-14-1
EINECS: 201-180-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 10 °C
Boiling Point 112 °C (1013 hPa)
Vapor Pressure 27.5 hPa (25 °C)
Density 1.26 g/cm3 (20 °C)
pH 0.5 (H2O)
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR