Dimethoxy(diisobutyl)silane for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diisobutyldimethoxysilane

M-Clarity™ quality level = MQ200

8.14720.0025 8.14720.0100
8.14720.0025EA 66.6 EUR
8.14720.0025 8.14720.0100
Dimethoxy(diisobutyl)silane for synthesis, Sigma-Aldrich®
Dimethoxy(diisobutyl)silane
Formule: C₁₀H₂₄O₂Si
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00236510
CAS nummer: 17980-32-4
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 13.4 mg/l (25 °C) Hydrolysis
Molar Mass 204.38 g/mol
Boiling Point 188 °C (1025 hPa)
Vapor Pressure 0.75 hPa (25 °C)
Flash Point 62 °C
Density 0.863 g/cm3 (20 °C)
Ignition Point 255 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR