Octamethylcyclotetrasiloxaan for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.14750.0025 8.14750.0100 8.14750.0250
8.14750.0025EA 34.2 EUR
8.14750.0025 8.14750.0100 8.14750.0250
Octamethylcyclotetrasiloxaan for synthesis, Sigma-Aldrich®
Octamethylcyclotetrasiloxaan
Formule: [-Si(CH₃)₂O-]₄
Kookpunt: 175 °C (1013 hPa)
Smeltpunt: 18 °C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 51 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003269
CAS nummer: 556-67-2
EINECS: 209-136-7
UN: 1993
ADR: 3,II
Merck Index: 12,06843

Order Now

Specification Test Results

Solubility  - 0.001 g/l (25 °C)
Melting Point 18 °C
Molar Mass 296.62 g/mol
Boiling Point 176 °C
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 51 °C
Explosion Limit 0.4 - 11.7 %(V)
Density 0.95 g/cm3
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR