o-Dichlorobenzene : Phenol mixture (50:50) (w/w) for synthesis, for measuring the viscosity acc. to DIN 53728, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.14765.2500
8.14765.2500EA 560 EUR
8.14765.2500
o-Dichlorobenzene : Phenol mixture (50:50) (w/w) for synthesis, for measuring the viscosity acc. to DIN 53728, Sigma-Aldrich®
o-Dichlorobenzene : Phenol mixture
Bewaar temperatuur: Kamertemperatuur
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Assay of phenole (GC, weight%) 48.0 - 52.0 %
Water (K. F.) ≤ 200 ppm
Assay of dichloro benzene (GC, weight%) 48.0 - 52.0 %
Colourless liquid passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR