Isopropyl myristate Ph. Eur., NF, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

8.17066.2500
8.17066.2500EA 560 EUR
8.17066.2500
Isopropyl myristate Ph. Eur., NF, SAFC®
Isopropyl myristate
Formule: C₁₇H₃₄O₂
MW: 270.46 g/mol
Kookpunt: 320
Smeltpunt: 0…1 °C
Dichtheid: 0.852 g/cm³ (20 °C)
Vlampunt: 150
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008982
CAS nummer: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR