(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.18316.0001 8.18316.0005
8.18316.0001EA 128 EUR
8.18316.0001 8.18316.0005
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine for synthesis, Sigma-Aldrich®
(2S,5SR)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropyl-5-methylpyrazine
Formule: C₁₀H₁₈N₂O₂
CAS nummer: 116907-51-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 198.26 g/mol
Boiling Point 103 - 104 °C (21 hPa)
Density 1.00 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR