Allyl cyanoacetate, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.18559.0050 8.18559.1000
8.18559.0050EA 230 EUR
8.18559.0050 8.18559.1000
Allyl cyanoacetate, Sigma-Aldrich®
Allyl cyanoacetate
Formule: C₆H₇NO₂
Smeltpunt: –40 °C
Dichtheid: 1,07 g/cm³ (20 °C)
Vlampunt: 91 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013817
CAS nummer: 13361-32-5
EINECS: 236-423-4
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -40.0 °C
Molar Mass 125.13 g/mol
Boiling Point 89 - 90 °C (5 hPa)
Vapor Pressure 0.05 hPa
Flash Point 91 °C
Refractive Index 1.4393 (28 °C, 589 nm)
Density 1.072 g/cm3 (20 °C)
Synonyms Cyanoacetic acid allyl ester
Hill Formula C6H7NO2
Chemical Formula NCCH2COOCH2CH=CH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR