(Methoxymethyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.18580.0050
8.18580.0050EA 92.8 EUR
8.18580.0050
(Methoxymethyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®
(Methoxymethyl)triphenylphosphonium chloride
Formule: C₂₀H₂₀ClOP
Smeltpunt: 190 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011800
CAS nummer: 4009-98-7
EINECS: 223-664-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) soluble,(decomposition)
Melting Point 202 °C
Molar Mass 342.8 g/mol
Bulk Density 450 - 500 kg/m3
Boiling Point 413 °C
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 250 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR