Zinc bromide for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Zinc (II) bromide

M-Clarity™ quality level = MQ200

8.18631.0005 8.18631.0250 8.18631.1000 8.18631.9025
8.18631.0005EA 29 EUR
8.18631.0005 8.18631.0250 8.18631.1000 8.18631.9025
Zinc bromide for synthesis, Sigma-Aldrich®
Zinc bromide
Formule: Br₂Zn
MW: 225.2 g/mol
Kookpunt: 670…680 °C (1013 hPa)
Smeltpunt: 394
Dichtheid: 4.22 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011294
CAS nummer: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8 III
Merck Index: 13,10182

Order Now

Specification Test Results

Solubility 4470 g/l (20 °C)
Melting Point 394 °C
Molar Mass 225.2 g/mol
Boiling Point 697 °C (1013 mbar)
Density 4.2 g/cm3 (20 °C)
pH 4 (H2O, 20 °C) (saturated solution)
Hill Formula Br2Zn
Chemical Formula ZnBr2

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR