Dimethyl pyrocarbonate for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.18758.0100
8.18758.0100EA 397 EUR
8.18758.0100
Dimethyl pyrocarbonate for synthesis, Sigma-Aldrich®
Dimethyl pyrocarbonate
Formule: C₄H₆O₅
Kookpunt: ∼172 °C (1013 hPa)
Smeltpunt: 17 °C
Dichtheid: 1,25 g/cm³ (20 °C)
Vlampunt: 84 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008419
CAS nummer: 4525-33-1
EINECS: 224-859-8
UN: 2927
ADR: 6.1 (8),II

Order Now

Specification Test Results

Solubility 35 g/l (20 °C) (decomposition)
Melting Point 15 - 17 °C
Molar Mass 134.09 g/mol
Boiling Point 172 °C (1013 hPa) decomposes
Vapor Pressure 0.7 hPa (20 °C)
Flash Point 85 °C
Refractive Index 1.395 (20 °C, 589 nm)
Explosion Limit 3 - 29.9 %(V)
Density 1.25 g/cm3 (20 °C)
Ignition Point 465 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR