Trimethyl orthoacetate, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.18804.0100 8.18804.0500
8.18804.0100EA 65.7 EUR
8.18804.0100 8.18804.0500
Trimethyl orthoacetate, Sigma-Aldrich®
Trimethyl orthoacetate
Formule: C₅H₁₂O₃
Kookpunt: 109 °C (1013 hPa)
Smeltpunt: –58 °C
Dichtheid: 0,944 g/cm³ (20 °C)
Vlampunt: 15 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008477
CAS nummer: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 120.14 g/mol
Boiling Point 108 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Flash Point 15 °C
Refractive Index 1.3859 (25 °C, 589 nm)
Density 0.96 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR