N-(3-(Trimethoxysilyl)propyl)ethylenediamine for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.19172.0100 8.19172.0500 8.19172.0025
8.19172.0100EA 61.9 EUR
8.19172.0100 8.19172.0500 8.19172.0025
N-(3-(Trimethoxysilyl)propyl)ethylenediamine for synthesis, Sigma-Aldrich®
N-(3-(Trimethoxysilyl)propyl)ethylenediamine
Formule: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
Kookpunt: 261…263 °C (1013 hPa)
Smeltpunt: ∼ 0 °C
Dichtheid: 1,01 g/cm³ (20 °C)
Vlampunt: 136 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008173
CAS nummer: 1760-24-3
EINECS: 217-164-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble,Hydrolysis
Molar Mass 222.36 g/mol
Boiling Point 261 - 263 °C (1013 hPa)
Vapor Pressure 1.5 hPa (20 °C)
Flash Point 136 °C
Density 1.02 g/cm3 (20 °C)
pH 10 (10 g/l, H2O, 20 °C)
Ignition Point 300 °C
Hill Formula C8H22N2O3Si


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR