Azelaic acid ≥90% (by GC), Technical Grade for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid

M-Clarity™ quality level = MQ200

8.20116.0100 8.20116.1000 8.20116.9050 8.20116.9025
8.20116.0100EA 41.6 EUR
8.20116.0100 8.20116.1000 8.20116.9050 8.20116.9025
Azelaic acid ≥90% (by GC), Technical Grade for synthesis, Sigma-Aldrich®
Azelaic acid
Formule: C₉H₁₆O₄
MW: 188.22 g/mol
Kookpunt: 371
Smeltpunt: 106
Dichtheid: 1.0287 g/cm³ (20 °C)
Vlampunt: 215
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004432
CAS nummer: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908

Order Now

Specification Test Results

Solubility 2.4 g/l (20 °C)
Melting Point 107 °C
Molar Mass 188.22 g/mol
Boiling Point 500 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 215 °C
Density 1.029 g/cm3 (20 °C)
pH 3.5 (1 g/l, H2O)
Synonyms Nonanedioic acid
Hill Formula C9H16O4
Chemical Formula HOOC(CH2)7COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR