4-tert-Butylbenzoic acid for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-(tert-Butyl)benzoic acid

M-Clarity™ quality level = MQ200

8.20238.0100 8.20238.1000 8.20238.9025
8.20238.0100EA 38.7 EUR
8.20238.0100 8.20238.1000 8.20238.9025
4-tert-Butylbenzoic acid for synthesis, Sigma-Aldrich®
4-tert-Butylbenzoic acid
Formule: C₁₁H₁₄O₂
MW: 178.23 g/mol
Kookpunt: 283
Smeltpunt: 165…167 °C
Dichtheid: 0.6 g/cm³ (20 °C)
Vlampunt: 180
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002563
CAS nummer: 98-73-7
EINECS: 202-696-3
UN: 2811

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 165 - 167 °C
Molar Mass 178.23 g/mol
Bulk Density 340 kg/m3
Boiling Point 280 °C (decomposition)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 180 °C
Density 0.6 g/cm3 (20 °C)
pH 3.9 (H2O) (saturated solution)
Ignition Point 510 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR