Dibutyl disulphide for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.20242.0025 8.20242.0100 8.20242.0010
8.20242.0025EA 64 EUR
8.20242.0025 8.20242.0100 8.20242.0010
Dibutyl disulphide for synthesis, Sigma-Aldrich®
Dibutyl disulphide
Formule: C₈H₁₈S₂
Kookpunt: 229 °C (1013 hPa)
Smeltpunt: –94 °C
Dichtheid: 0,9371 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009467
CAS nummer: 629-45-8
EINECS: 211-091-3

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -94 °C
Molar Mass 178.35 g/mol
Boiling Point 229 - 233 °C
Flash Point 93 °C
Refractive Index 1.492 (20 °C, 589 nm)
Explosion Limit 0.8 %(V)
Density 0.94 g/cm3 (20 °C)
Synonyms Butyl disulfide
Hill Formula C8H18S2
Chemical Formula CH3(CH2)3SS(CH2)3CH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR