2-tert-Butyl-4-methoxyphenol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.20246.0100 8.20246.9025 8.20246.0500 8.20246.0025 8.20246.5000
8.20246.0100EA 67.8 EUR
8.20246.0100 8.20246.9025 8.20246.0500 8.20246.0025 8.20246.5000
2-tert-Butyl-4-methoxyphenol for synthesis, Sigma-Aldrich®
2-tert-Butyl-4-methoxyphenol
Formule: C₁₁H₁₆O₂
Kookpunt: 267…269 °C (1013 hPa)
Smeltpunt: 60…61 °C
Dichtheid: 1,0587 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040484
CAS nummer: 121-00-6
EINECS: 204-442-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 48 - 63 °C
Molar Mass 180.25 g/mol
Boiling Point 263 °C (1013 hPa)
Vapor Pressure 1.33 hPa (105 °C)
Flash Point 130 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR