2,4-Dichloorfenoxyazijnzuur, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.20451.0005 8.20451.0250 8.20451.1000 8.20451.9020
8.20451.0005EA 26.5 EUR
8.20451.0005 8.20451.0250 8.20451.1000 8.20451.9020
2,4-Dichloorfenoxyazijnzuur, Sigma-Aldrich®
2,4-Dichloorfenoxyazijnzuur
Formule: (C₆H₃Cl₂)OCH₂COOH
Kookpunt: 346 °C (1013 hPa)
Smeltpunt: 137…139 °C
Dichtheid: 1,563 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004300
CAS nummer: 94-75-7
EINECS: 202-361-1
Merck Index: 13,02825

Order Now

Specification Test Results

Solubility 0.3 g/l (25 °C)
Melting Point 134 - 137 °C
Molar Mass 221.04 g/mol
Bulk Density 720 kg/m3
Boiling Point 160 °C (0.53 hPa)
Density 1.42 g/cm3 (25 °C)
Synonyms 2,4 D-acid
Hill Formula C8H6Cl2O3
Chemical Formula 2,4-(Cl)2C6H3OCH2COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR