Allyl acetate for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Acetic acid allyl ester

M-Clarity™ quality level = MQ200

8.20554.0100 8.20554.0500
8.20554.0100EA 79 EUR
8.20554.0100 8.20554.0500
Allyl acetate for synthesis, Sigma-Aldrich®
Allyl acetate
Formule: C₅H₈O₂
MW: 100.12 g/mol
Kookpunt: 103
Smeltpunt: –96 °C
Dichtheid: 0.93 g/cm³ (20 °C)
Vlampunt: 6
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008721
CAS nummer: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3 (6.1) II
Merck Index: 12,00293

Order Now

Specification Test Results

Solubility 28 g/l (20 °C)
Melting Point -96 °C
Molar Mass 100.11 g/mol
Boiling Point 103 - 104 °C (1013 mbar)
Vapor Pressure 27.2 hPa (20 °C)
Flash Point 6 °C
Refractive Index 1.4042 (20 °C, 589 nm)
Explosion Limit 2.1 - 13.0 %(V)
Density 0.93 g/cm3 (20 °C)
Ignition Point 374 °C
Synonyms Acetic acid allyl ester
Hill Formula C5H8O2
Chemical Formula CH3COOCH2CH=CH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR