5-(Hydroxymethyl)-2-furaldehyde zS, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural

MQ200

8.20678.0001 8.20678.0005 8.20678.0200 8.20678.8100
8.20678.0001EA 97.2 EUR
8.20678.0001 8.20678.0005 8.20678.0200 8.20678.8100
5-(Hydroxymethyl)-2-furaldehyde zS, Sigma-Aldrich®
5-(Hydroxymethyl)-2-furaldehyde
Formule: C₆H₆O₃
MW: 126.11 g/mol
Kookpunt: 292
Smeltpunt: 27…30 °C
Dichtheid: 1.24 g/cm³ (20 °C)
Vlampunt: 79
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003234
CAS nummer: 67-47-0
EINECS: 200-654-9
Merck Index: 13,04857

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 27 - 30 °C
Molar Mass 126.11 g/mol
Boiling Point 154 - 155 °C (16 hPa)
Flash Point 79 °C
Refractive Index 1.5627 (20 °C, 589 nm)
Synonyms 5-(Hydroxymethyl)furfurol
Hill Formula C6H6O3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR