Ethyl iodoacetate zS

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

8.20733.1000
8.20733.1000EA 0 EUR
8.20733.1000
Ethyl iodoacetate zS
Ethyl iodoacetate
Formule: C₄H₇IO₂
MW: 214 g/mol
Kookpunt: 179
Dichtheid: 1.81 g/cm³ (20 °C)
MDL Nummer: MFCD00001081
CAS nummer: 623-48-3
EINECS: 210-796-3
UN: 2927
ADR: 6.1 II

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 214 g/mol
Boiling Point 179 °C (1013 hPa)
Vapor Pressure 0.7 hPa (20 °C)
Flash Point 76 °C
Refractive Index 1.5045 (20 °C, 589 nm)
Density 1.81 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR