Oleyl alcohol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.20923.0100 8.20923.1000 8.20923.2500 8.20923.9050
8.20923.0100EA 29.6 EUR
8.20923.0100 8.20923.1000 8.20923.2500 8.20923.9050
Oleyl alcohol for synthesis, Sigma-Aldrich®
Oleyl alcohol
Formule: C₁₈H₃₆O
Kookpunt: 333 °C (1013 hPa)
Smeltpunt: 13…19 °C
Dichtheid: 0,85 g/cm³ (20 °C)
MDL Nummer: MFCD00002993
CAS nummer: 143-28-2
EINECS: 205-597-3
Merck Index: 12,06968

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 13 - 19 °C
Molar Mass 268.48 g/mol
Boiling Point 330 - 360 °C (1013 hPa)
Vapor Pressure 1013 hPa (335 °C)
Flash Point 112 °C
Refractive Index 1.461 (20 °C, 589 nm)
Density 0.85 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR