1-Octanol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.20931.0100 8.20931.1000 8.20931.2500 8.20931.9025 8.20931.9165 8.20931.0025 8.20931.9170
8.20931.0100EA 18.6 EUR
8.20931.0100 8.20931.1000 8.20931.2500 8.20931.9025 8.20931.9165 8.20931.0025 8.20931.9170
1-Octanol for synthesis, Sigma-Aldrich®
1-Octanol
Formule: H₃C(CH₂)₆CH₂OH
Kookpunt: 195 °C (1013 hPa)
Smeltpunt: –16 °C
Dichtheid: 0,8246 g/cm³ (20 °C)
Vlampunt: 81 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002988
CAS nummer: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point -16 °C
Molar Mass 130.23 g/mol
Boiling Point 195 °C (1013 hPa)
Vapor Pressure 0.031 hPa (20 °C)
Flash Point 90 °C
Explosion Limit 0.8 %(V)
Density 0.83 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 270 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR