Phenylacetic acid for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.20993.0250 8.20993.1000
8.20993.0250EA 36.4 EUR
8.20993.0250 8.20993.1000
Phenylacetic acid for synthesis, Sigma-Aldrich®
Phenylacetic acid
Formule: C₈H₈O₂
MW: 136.15 g/mol
Kookpunt: 265
Smeltpunt: 74…77 °C
Dichtheid: 1.081 g/cm³ (20 °C)
Vlampunt: 132
MDL Nummer: MFCD00004313
CAS nummer: 103-82-2
EINECS: 203-148-6
Merck Index: 12,07422

Order Now

Specification Test Results

Solubility 1.58 g/l (20 °C)
Melting Point 74 - 77 °C
Molar Mass 136.15 g/mol
Boiling Point 265 °C (1013 hPa)
Vapor Pressure 1.33 hPa (97 °C)
Flash Point 132 °C
Density 1.091 g/cm3 (97 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR