N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.21102.0010 8.21102.0025 8.21102.0002
8.21102.0010EA 135 EUR
8.21102.0010 8.21102.0025 8.21102.0002
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride for synthesis, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formule: C₁₀H₁₈Cl₂N₂
Smeltpunt: 227…230 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012482
CAS nummer: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e955f6b2a1434ddba239f533c6611583219f8904&vid=13fb110b013db70c026f8eefeee7f08fa2568c0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 227 - 230 °C (decomposition)
Molar Mass 237.17 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR