Tetraethylammonium bromide for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tetrylammonium bromide

M-Clarity™ quality level = MQ200

8.22147.0250 8.22147.1000 8.22147.0050
8.22147.0250EA 59.9 EUR
8.22147.0250 8.22147.1000 8.22147.0050
Tetraethylammonium bromide for synthesis, Sigma-Aldrich®
Tetraethylammonium bromide
Formule: C₈H₂₀BrN
MW: 210.16 g/mol
Smeltpunt: > 285 °C
Dichtheid: 1.43 g/cm³ (20 °C)
MDL Nummer: MFCD00011825
CAS nummer: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Order Now

Specification Test Results

Solubility 2795 g/l (25 °C)
Melting Point 285 °C
Molar Mass 210.16 g/mol
Density 1.397 g/cm3 (20 °C)
pH 6.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR