Isoamyl alcohol (mixture of isomers), EMPLURA®, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.22255.1000 8.22255.2500
8.22255.1000EA 55.1 EUR
8.22255.1000 8.22255.2500
Isoamyl alcohol (mixture of isomers), EMPLURA®, Supelco®
Isoamyl alcohol (mixture of isomers)
Formule: C₅H₁₂O
Kookpunt: 131 °C (1013 hPa)
Smeltpunt: –117 °C
Dichtheid: 0,81 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002977
CAS nummer: 30899-19-5
EINECS: 250-378-8
UN: 1105
ADR: 3,III

Order Now

Specification Test Results

Assay isomers (GC, area%) ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.808 - 0.812
Water (K. F.) ≤ 0.30 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR