Natriumsulfaat for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

M-Clarity™ quality level = MQ200

8.22286.5000
8.22286.5000EA 122 EUR
8.22286.5000
Natriumsulfaat for synthesis, Sigma-Aldrich®
Natriumsulfaat
Formule: Na₂SO₄
Kookpunt: 1429 °C (1013 hPa)
Smeltpunt: 888 °C
Dichtheid: 2,7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 44.55 g/l (20 °C)
Melting Point 888 °C
Molar Mass 142.04 g/mol
Bulk Density 1400 - 1600 kg/m3
Density 2.70 g/cm3 (20 °C)
pH 4 - 7 (200 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR