Citraconic anhydride for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.41321.0100
8.41321.0100EA 105 EUR
8.41321.0100
Citraconic anhydride for synthesis, Sigma-Aldrich®
Citraconic anhydride
Formule: C₅H₄O₃
Kookpunt: 213 °C (1013 hPa)
Smeltpunt: 7…8 °C
Dichtheid: 1,243 g/cm³ (20 °C)
Vlampunt: 101 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005522
CAS nummer: 616-02-4
EINECS: 210-459-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Melting Point 7 - 8 °C
Molar Mass 112.08 g/mol
Boiling Point 213 - 214 °C (1013 hPa)
Vapor Pressure 1.3 hPa (47 °C)
Flash Point 101 °C
Density 1.243 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR