Pentafluorobenzoyl chloride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC

M-Clarity™ quality level = MQ200

8.41331.0005 8.41331.0025
8.41331.0005EA 80 EUR
8.41331.0005 8.41331.0025
Pentafluorobenzoyl chloride ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Pentafluorobenzoyl chloride
Formule: C₇ClF₅O
MW: 230.52 g/mol
Kookpunt: 158
Dichtheid: 1.6 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000657
CAS nummer: 2251-50-5
EINECS: 218-842-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Assay (GC, area%) ≥98.0%
Identity (IR) Passes test
Boiling Point 158 - 159 °C (1013 hPa)
Density 1.664 g/cm3 (20 °C)
pH (H2O, 20 °C) acidic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR