2,4-Diamino-6-hydroxypyrimidine for synthesis

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.41789.0050
8.41789.0050EA 102 EUR
8.41789.0050
2,4-Diamino-6-hydroxypyrimidine for synthesis
2,4-Diamino-6-hydroxypyrimidine
Formule: C₄H₆N₄O
Kookpunt: 373 °C (1013 hPa)
Smeltpunt: 285…286 °C
MDL Nummer: MFCD00006098
CAS nummer: 56-06-4
EINECS: 200-254-4
Merck Index: 13,03001

Order Now

Specification Test Results

Melting Point 285 - 286 °C
Molar Mass 126.11 g/mol
Hill Formula C4H6N4O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR