1,1,3,3-Tetramethyl-2-thiourea for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: N,N,N',N'-Tetramethylthiourea, Tetramethyl-2-thiourea, Tetramethylthiourea

M-Clarity™ quality level = MQ200

8.41854.9999
8.41854.9999EA 0 EUR
8.41854.9999
1,1,3,3-Tetramethyl-2-thiourea for synthesis, Sigma-Aldrich®
1,1,3,3-Tetramethyl-2-thiourea
Formule: C₅H₁₂N₂S
MW: 132.23 g/mol
Kookpunt: 245
Smeltpunt: 74…76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008324
CAS nummer: 2782-91-4
EINECS: 220-488-0

Order Now

Specification Test Results

Solubility 32 g/l (20 °C)
Melting Point 75 - 78 °C
Molar Mass 132.22 g/mol
Boiling Point 245 °C (1013 hPa)
Vapor Pressure 1.9 hPa (25 °C)
Flash Point 100 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR