Benzyltrimethylammonium hydroxide 40% in aqueous solution

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8.45025.5000 8.45025.0100
8.45025.5000EA 0 EUR
8.45025.5000 8.45025.0100
Benzyltrimethylammonium hydroxide 40% in aqueous solution
Benzyltrimethylammonium hydroxide
Formule: C₁₀H₁₇NO
MDL Nummer: MFCD00008281
CAS nummer: 100-85-6
EINECS: 202-895-5
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR