Natriumsulfaat, anhydrous 99,0 Dry_basis USP, Macron Fine Chemicals™

Leverancier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

M.O.C. Category R

8028-06EA 173.31 EUR
8028-06 8028-88 8028-12 8028-65 8028-F9 8028-W3 MACR8028-20
Natriumsulfaat, anhydrous 99,0 Dry_basis USP, Macron Fine Chemicals™
Natriumsulfaat

No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.

Formule: Na₂SO₄
MW: 142.04 g/mol
Kookpunt: 1429 °C (1013 hPa)
Smeltpunt: 888 °C
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical = 99.0 %
USP - Assay (Na₂SO₄) (dried basis) ≥ 99.0 %
USP - Acidity or Alkalinity (mL) ≤ 0.50
USP - Chloride (Cl) ≤ 0.005 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤ 0.01 %
USP - Loss on Drying at 105°C ≤ 0.5 %
Thru U.S. No. 50 Sieve, For Information Only = 0.5 %
No Class 1,2,3 or other solvents are used or produced in the For Information Only
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.
Appearance - Colorless to slightly colored crystalline solid.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR