Anti-static cleanroom tapes, NovaStat™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ASTPC-272_U ASTPC-3436
819-0073EA 0 EUR
819-0073 819-0017
Anti-static cleanroom tapes, NovaStat™
Tape
These anti-static cleanroom tapes are made of cellophane with rubber based adhesive and are specially designed for use in ESD-sensitive environments.
  • Suitable for sealing and packaging
  • Will not create a static charge
  • Wound on a 3" plastic core
Colour: Amber transparent