Bewaringssysteem, Cryoinstant

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

822075ZAEA 186 EUR
822075ZA 822070ZA 822071ZA 822072ZA 822073ZA 822074ZA 820006ZA
Bewaringssysteem, Cryoinstant
Testsystemen voor microbiologie Micro-organisme testen

Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.


  • Quick and easy to use
  • Colour coded
  • Storage down to –70 °C
  • Quality controlled for fertility and sterility


Verpakking: Cryoinstant is packed in packs of 50 vials of 2 ml.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR