Tolueen ≥99.7%, PESTINORM® for pesticide residue analysis

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Methylbenzene, Phenylmethane, FM approval

Filtered through a 0.2 µm filter, packaged under nitrogen.

83664.320EA 96.1 EUR
83664.320 83664.400
Tolueen ≥99.7%, PESTINORM® for pesticide residue analysis
Tolueen
Formule: C₆H₅CH₃
Kookpunt: 110,6 °C (1013 hPa)
Smeltpunt: –95 °C
Dichtheid: 0,867 g/cm³ (20 °C)
Vlampunt: 4 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
Bereik: 01-2119471310-51
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.7 %
Evaporation residue Max. 5 ppm
Water Max. 0.03 %
Lindane (GC/ECD) ≤ 5 ng/l Passes test
Pesticide analysis (Lindane/ECD) Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR