Salpeterzuur fuming 98 Titrimetric, Purum p.a. analytical reagent

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Nitric acid solution

coated,shock-leak-protected glass bottle

84392-500ML
84392-500MLEA 174 EUR
84392-500ML
Salpeterzuur fuming 98 Titrimetric, Purum p.a. analytical reagent
Salpeterzuur
Formule: HNO₃
MW: 63.01 g/mol
Kookpunt: 86 °C
Smeltpunt: –42 °C
Dichtheid: 1.52 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2032
ADR: 8 I
Bereik: 01-2119487297-23
Merck Index: 13,06610

Order Now

Specification Test Results

Assay Min. 98 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 150 ppm
Chloride (Cl) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR