Ethylacetaat ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Gevaar
Filtered through a 0.2 µm filter, packaged under nitrogen.
84532.550EA 2244 EUR
84532.550 84532.460 84532.950
Ethylacetaat ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Ethylacetaat
Formule: CH₃COOC₂H₅
Kookpunt: 77,1 °C (1013 hPa)
Smeltpunt: –83 °C
Dichtheid: 0,902 g/cm³ (20 °C)
Vlampunt: –4 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009171
CAS nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
Bereik: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (calculated on anhydrous) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 0.0005 %
Water Max. 0.02 %