Azijnzuur 99% 99, HiPerSolv CHROMANORM® LC-MS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

Nitrogen_filled

84874.180EA 34.3 EUR
84874.180 84874.260 84874.290
Azijnzuur 99% 99, HiPerSolv CHROMANORM® LC-MS
Azijnzuur
Formule: H₃CCOOH
MW: 60.05 g/mol
Kookpunt: 118 °C (1013 hPa)
Smeltpunt: 17 °C
Dichtheid: 1.05 g/cm³ (20 °C)
Vlampunt: 38.5 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036152
CAS nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
Bereik: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8%
Evaporation residue Max. 0.0001%
Water Max. 0.1%
Calcium (0.1% Aqueous solution) Max. 0.2 ppm
Potassium (0.1% Aqueous solution) Max. 0.2 ppm
Magnesium (0.1% Aqueous solution) Max. 0.2 ppm
Sodium (0.1% Aqueous solution) Max. 0.5 ppm
Transmittance (255 nm) Min 10%
Transmittance (260 nm) Min 50%
Transmittance (270 nm) Min 80%
Transmittance (280 nm) Min 95%
Transmittance (300 nm) Min 99%
Suitability for LC-MS (0.1% aqueous solution) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR